Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-5-(trifluoromethyl)pyridine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

24181-0250 24181-1000
TS24181-0250EA 39 USD InStock
TS24181-0250 TS24181-1000
2,3-Dichloro-5-(trifluoromethyl)pyridine 97%
2,3-Dichloro-5-(trifluoromethyl)pyridine


Formula: C₆H₂Cl₂F₃N
Boiling Pt: 176 °C (760 mmHg)
Melting Pt: 8…9 °C
Density: 1.549 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00042243
CAS Number: 69045-84-7
EINECS: 410-340-5
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4745 to 1.4765 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty