Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-hydroxy-8-mercaptopurine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

29523-0050 29523-0250
TS29523-0050EA 80 USD InStock
TS29523-0050 TS29523-0250
2-Amino-6-hydroxy-8-mercaptopurine 97%
2-Amino-6-hydroxy-8-mercaptopurine


Formula: C₅H₅N₅OS
MW: 183.189 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00075623
CAS Number: 28128-40-7
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty