Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-benzoxazolethiol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

29730-0050 29730-0250
TS29730-0050EA 60 USD InStock
TS29730-0050 TS29730-0250
6-Chloro-2-benzoxazolethiol 99%
6-Chloro-2-benzoxazolethiol


Formula: C₇H₄ClNOS
MW: 185.625 g/mol
Melting Pt: 229…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00800495
CAS Number: 22876-20-6
Order Now
Specification Test Results
Appearance Yellow to beige Crystalline powder or needles
Infrared spectrum Conforms
Melting point 229 to 232 °C
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty