Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dimethyldibenzothiophene 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30219-0010 30219-0050
TS30219-0010EA 126 USD InStock
TS30219-0010 TS30219-0050
4,6-Dimethyldibenzothiophene 95%
4,6-Dimethyldibenzothiophene


Formula: C₁₄H₁₂S
Melting Pt: 147…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00216264
CAS Number: 1207-12-1
Order Now
Specification Test Results
Appearance Light yellow to beige to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 150 to 156 °C
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty