Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thymoquinone

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30507-0010 30507-0050
TS30507-0010EA 48 USD InStock
TS30507-0010 TS30507-0050
Thymoquinone
Thymoquinone


Formula: C₁₀H₁₂O₂
Boiling Pt: 230…232 °C (760 mmHg)
Melting Pt: 43…44 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00001602
CAS Number: 490-91-5
Order Now
Specification Test Results
Appearance Yellow to orange to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 44 to 47 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty