Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1,3-benzodioxole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

30768-0250 30768-1000
TS30768-0250EA 99 USD InStock
TS30768-0250 TS30768-1000
5-Bromo-1,3-benzodioxole 97%
5-Bromo-1,3-benzodioxole


Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 85…86 °C (1 mmHg)
Density: 1.66 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005821
CAS Number: 2635-13-4
EINECS: 220-123-5
Order Now
Specification Test Results
Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5820 to 1.5850 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty