Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoquinoline 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

32503-1000 32503-5000
TS32503-1000EA 75 USD InStock
TS32503-1000 TS32503-5000
Isoquinoline 97%
Isoquinoline


Formula: C₉H₇N
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 24…28 °C
Density: 1.09 g/cm³
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006898
CAS Number: 119-65-3
EINECS: 204-341-8
UN: 2811
ADR: 6.1,II
Merck Index: 13,05242
Order Now
Specification Test Results
Appearance Colorless to white to yellow to light brown Fused solid or liquid
Infrared spectrum Conforms
Melting point 24 to 28 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty