Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydropyran 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

32588-1000 32588-5000
TS32588-1000EA 69 USD InStock
TS32588-1000 TS32588-5000
Tetrahydropyran 99%
Tetrahydropyran


Formula: C₅H₁₀O
Boiling Pt: 88 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.88 g/cm³
Flash Pt: –15 °C
MDL Number: MFCD00006585
CAS Number: 142-68-7
EINECS: 205-552-8
UN: 3271
ADR: 3,II
Merck Index: 12,09356
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty