Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Thienyl)propanoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

33011-0010 33011-0050
TS33011-0010EA 33 USD InStock
TS33011-0010 TS33011-0050
3-(2-Thienyl)propanoic acid 98%
3-(2-Thienyl)propanoic acid


Formula: C₇H₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD00047093
CAS Number: 5928-51-8
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to beige Crystals or needles
Infrared spectrum Conforms
Melting point 45 to 49 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty