Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dibromopyridine 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33013-0100 33013-0250 33013-1000
TS33013-0100EA 108 USD InStock
TS33013-0100 TS33013-0250 TS33013-1000
3,5-Dibromopyridine 98+%
3,5-Dibromopyridine


Formula: C₅H₃Br₂N
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 110…114 °C
Flash Pt: >115 °C
Storage Temperature: Ambient
MDL Number: MFCD00014634
CAS Number: 625-92-3
EINECS: 210-916-4
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to light beige Powder or crystals or needles
Infrared spectrum Conforms
Melting point 110 to 114 °C
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty