Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Allylimidazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

33517-0250 33517-1000
TS33517-0250EA 53 USD InStock
TS33517-0250 TS33517-1000
1-Allylimidazole 97%
1-Allylimidazole


Formula: C₆H₈N₂
Boiling Pt: 212…213 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00037894
CAS Number: 31410-01-2
EINECS: 250-618-1
UN: 3267
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5060 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty