Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-6-methoxypyrimidine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

33745-0050 33745-0250
TS33745-0050EA 92 USD InStock
TS33745-0050 TS33745-0250
4-Amino-6-methoxypyrimidine 97%
4-Amino-6-methoxypyrimidine


Formula: C₅H₇N₃O
MW: 125.1286 g/mol
Melting Pt: 154…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00129983
CAS Number: 696-45-7
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 154 to 159 °C
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty