Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thiopheneboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

34566-0010 34566-0050
TS34566-0010EA 32 USD InStock
TS34566-0010 TS34566-0050
3-Thiopheneboronic acid 97%
3-Thiopheneboronic acid


Formula: C₄H₅BO₂S
MW: 127.96
Melting Pt: 162…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151851
CAS Number: 6165-69-1
Order Now
Specification Test Results
Appearance White to beige to grey Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with NaOH 96.0 to 106.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty