Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Furfuryl glycidyl ether 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

34849-0050 34849-0250
TS34849-0050EA 71 USD InStock
TS34849-0050 TS34849-0250
Furfuryl glycidyl ether 97%
Furfuryl glycidyl ether


Formula: C₈H₁₀O₃
Boiling Pt: 103…104 °C (11 mmHg)
Density: 1.122 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00013348
CAS Number: 5380-87-0
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty