Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-chloromethyl-2-furancarboxylate 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

34973-0010 34973-0050 34973-0250
TS34973-0010EA 33 USD InStock
TS34973-0010 TS34973-0050 TS34973-0250
Ethyl-5-chloromethyl-2-furancarboxylate 95%
Ethyl-5-chloromethyl-2-furancarboxylate


Formula: C₈H₉ClO₃
Boiling Pt: 115 °C (760 mmHg)
Density: 1.225 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00173837
CAS Number: 2528-00-9
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5100 to 1.5130 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty