Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Aminopropyl)imidazole 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

35194-0500 35194-2500
TS35194-0500EA 53 USD InStock
TS35194-0500 TS35194-2500
1-(3-Aminopropyl)imidazole 98%
1-(3-Aminopropyl)imidazole


Formula: C₆H₁₁N₃
Melting Pt: –68 °C
Flash Pt: 154 °C
MDL Number: MFCD00009819
CAS Number: 5036-48-6
EINECS: 225-730-9
UN: 2735
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5170 to 1.5210 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty