Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Aminonicotinonitrile 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35762-0050 35762-0250
TS35762-0050EA 59 USD InStock
TS35762-0050 TS35762-0250
6-Aminonicotinonitrile 98%
6-Aminonicotinonitrile


Formula: C₆H₅N₃
Boiling Pt: 240…250 °C (15 mmHg)
Melting Pt: 159…165 °C
MDL Number: MFCD00128902
CAS Number: 4214-73-7
Order Now
Specification Test Results
Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 165 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty