Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Furan-2-boronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35939-0010 35939-0100
TS35939-0010EA 56 USD InStock
TS35939-0010 TS35939-0100
Furan-2-boronic acid 97%
Furan-2-boronic acid


Formula: C₄H₅BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00799544
CAS Number: 13331-23-2
Order Now
Specification Test Results
Appearance Light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty