Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Furan-3-boronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

35940-0025 35940-1000 35940-5000
TS35940-0025EA 160 USD InStock
TS35940-0025 TS35940-1000 TS35940-5000
Furan-3-boronic acid 97%
Furan-3-boronic acid


Formula: C₄H₅BO₃
MW: 111.89 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01319007
CAS Number: 55552-70-0
Order Now
Specification Test Results
Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty