Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloro-6-methylpyridine 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36080-0010 36080-0050
TS36080-0010EA 37 USD InStock
TS36080-0010 TS36080-0050
3-Bromo-2-chloro-6-methylpyridine 98+%
3-Bromo-2-chloro-6-methylpyridine


Formula: C₆H₅BrClN
MW: 206.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095218
CAS Number: 185017-72-5
Order Now
Specification Test Results
Appearance Yellow Low melting solid or liquid
Infrared spectrum Conforms
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty