Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-methyl-3-nitropyridine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36083-0010 36083-0050
TS36083-0010EA 66 USD InStock
TS36083-0010 TS36083-0050
2-Chloro-6-methyl-3-nitropyridine 98%
2-Chloro-6-methyl-3-nitropyridine


Formula: C₆H₅ClN₂O₂
MW: 172.57
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD03085820
CAS Number: 56057-19-3
UN: 2811
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 69 to 74 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty