Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-iodopyridine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36594-0050 36594-0250
TS36594-0050EA 46 USD InStock
TS36594-0050 TS36594-0250
5-Bromo-2-iodopyridine 98%
5-Bromo-2-iodopyridine


Formula: C₅H₃BrIN
Melting Pt: 112…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00691506
CAS Number: 223463-13-6
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty