Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyanophenylboronic acid pinacol ester 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36643-0010 36643-0050
TS36643-0010EA 47 USD InStock
TS36643-0010 TS36643-0050
4-Cyanophenylboronic acid pinacol ester 97%
4-Cyanophenylboronic acid pinacol ester


Formula: C₁₃H₁₆BNO₂
Melting Pt: 94…99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093897
CAS Number: 171364-82-2
Order Now
Specification Test Results
Appearance Light yellow to orange to tan Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 99 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty