Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxycarbazole 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36696-0010 36696-0050
TS36696-0010EA 36 USD InStock
TS36696-0010 TS36696-0050
4-Hydroxycarbazole 98+%
4-Hydroxycarbazole


Formula: C₁₂H₉NO
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD02178385
CAS Number: 52602-39-8
EINECS: 258-034-9
Order Now
Specification Test Results
Appearance Tan to dark brown Powder
Infrared spectrum Conforms
Melting point 163 to 168 °C
HPLC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty