Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-(Trifluoromethyl)phenyl)piperazine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

36712-0010 36712-0050
TS36712-0010EA 55 USD InStock
TS36712-0010 TS36712-0050
1-(4-(Trifluoromethyl)phenyl)piperazine 98%
1-(4-(Trifluoromethyl)phenyl)piperazine


Formula: C₁₁H₁₃F₃N₂
MW: 230.23 g/mol
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00040765
CAS Number: 30459-17-7
EINECS: 250-210-3
UN: 3259
ADR: 8 III
Order Now
Specification Test Results
Appearance Colorless to light yellow Crystals
Infrared spectrum Conforms
Melting point 85 to 90 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty