Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[2-(Morpholin-4-yl)ethyl]piperazine 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

36714-0010 36714-0050
TS36714-0010EA 71 USD InStock
TS36714-0010 TS36714-0050
1-[2-(Morpholin-4-yl)ethyl]piperazine 99%
1-[2-(Morpholin-4-yl)ethyl]piperazine


Formula: C₁₀H₂₁N₃O
MW: 199.298 g/mol
Boiling Pt: 82…84 °C (.5mbar)
Storage Temperature: Ambient
MDL Number: MFCD00191217
CAS Number: 4892-89-1
UN: 2735
ADR: 8 III
Order Now
Specification Test Results
Appearance White to light yellow Low melting crystals or viscous liquid
Infrared spectrum Conforms
GC ≥98.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty