Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-methyl-1H-pyrazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36803-0050 36803-0250 36803-1000
TS36803-0050EA 30 USD InStock
TS36803-0050 TS36803-0250 TS36803-1000
3-Amino-5-methyl-1H-pyrazole 97%
3-Amino-5-methyl-1H-pyrazole


Formula: C₄H₇N₃
Melting Pt: 41…43 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075180
CAS Number: 31230-17-8
Order Now
Specification Test Results
Appearance Light yellow to orange Solid or viscous liquid
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty