Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrrole-2-carbonitrile 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36835-0010 36835-0050
TS36835-0010EA 50 USD InStock
TS36835-0010 TS36835-0050
Pyrrole-2-carbonitrile 97%
Pyrrole-2-carbonitrile


Formula: C₅H₄N₂
Boiling Pt: 115…120 °C (10 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00234061
CAS Number: 4513-94-4
UN: 3276
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5510 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty