Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Aminoquinoline 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

36964-0010 36964-0050 36964-0250
TS36964-0010EA 43 USD InStock
TS36964-0010 TS36964-0050 TS36964-0250
6-Aminoquinoline 98%
6-Aminoquinoline


Formula: C₉H₈N₂
Boiling Pt: 192…195 °C (14 mmHg)
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00006803
CAS Number: 580-15-4
EINECS: 209-453-0
Order Now
Specification Test Results
Appearance Yellow to grey-green to brown Powder
Infrared spectrum Conforms
Melting point 115 to 119 °C
HPLC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty