Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxyindole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

37901-0010 37901-0050
TS37901-0010EA 181 USD InStock
TS37901-0010 TS37901-0050
6-Hydroxyindole 97%
6-Hydroxyindole


Formula: C₈H₇NO
Melting Pt: 124…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00152101
CAS Number: 2380-86-1
EINECS: 417-020-4
UN: 3077
Order Now
Specification Test Results
Appearance White to off-white Glistening crystalline powder (May darken on storage)
Infrared spectrum Conforms
Melting point 124 to 130 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty