Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Isopropylpiperazine 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

38481-0010 38481-0050
TS38481-0010EA 69 USD InStock
TS38481-0010 TS38481-0050
1-Isopropylpiperazine 98+%
1-Isopropylpiperazine


Formula: C₇H₁₆N₂
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00167971
CAS Number: 4318-42-7
UN: 2733
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4720 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty