Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-6-methylpyridine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39105-0010 39105-0100
TS39105-0010EA 17 USD InStock
TS39105-0010 TS39105-0100
2-Bromo-6-methylpyridine 97%
2-Bromo-6-methylpyridine


Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 102…103 °C (20 mmHg)
Density: 1.512 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00040743
CAS Number: 5315-25-3
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5620 to 1.5640 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty