Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromothiazole 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39710-0010 39710-0050
TS39710-0010EA 45 USD InStock
TS39710-0010 TS39710-0050
2,5-Dibromothiazole 97%
2,5-Dibromothiazole


Formula: C₃HBr₂NS
Melting Pt: 45…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00016891
CAS Number: 4175-78-4
Order Now
Specification Test Results
Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 45 to 50 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty