Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-pyridineboronic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39777-0010 39777-0050
TS39777-0010EA 51 USD InStock
TS39777-0010 TS39777-0050
2-Chloro-5-pyridineboronic acid 97%
2-Chloro-5-pyridineboronic acid


Formula: C₅H₅BClNO₂
Melting Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD03094998
CAS Number: 444120-91-6
Order Now
Specification Test Results
Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 161 to 167 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty