Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azetidine hydrochloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

39873-0010 39873-0050
TS39873-0010EA 85 USD InStock
TS39873-0010 TS39873-0050
Azetidine hydrochloride 97%
Azetidine hydrochloride


Formula: C₃H₈ClN
Melting Pt: >300 °C
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00191762
CAS Number: 36520-39-5
Order Now
Specification Test Results
Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty