Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Bithiophene 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

40269-0025 40269-0200
TS40269-0025EA 31 USD InStock
TS40269-0025 TS40269-0200
2,2'-Bithiophene 97%
2,2'-Bithiophene


Formula: C₈H₆S₂
Melting Pt: 30…35 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005414
CAS Number: 492-97-7
EINECS: 207-767-2
Order Now
Specification Test Results
Appearance Off-white to dark green to blue Crystals or low melting crystalline mass
Infrared spectrum Conforms
Melting point 30 to 35 °C
GC ≥96.0 %
Specific optical rotation +42° to +48° (20 °C, 589 nm) (c=3, H2O)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty