Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Epoxytetrahydrofuran 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42553-0010 42553-0100 42553-0500
TS42553-0010EA 90 USD InStock
TS42553-0010 TS42553-0100 TS42553-0500
3,4-Epoxytetrahydrofuran 96%
3,4-Epoxytetrahydrofuran


Formula: C₄H₆O₂
MW: 86.09 g/mol
Density: 1.237 g/cm³
Flash Pt: >38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00800639
CAS Number: 285-69-8
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4490 (20°C, 589 nm)
GC ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty