Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-thiophenecarboxylic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42684-0010 42684-0050
TS42684-0010EA 88 USD InStock
TS42684-0010 TS42684-0050
2-Bromo-3-thiophenecarboxylic acid 97%
2-Bromo-3-thiophenecarboxylic acid


Formula: C₅H₃BrO₂S
MW: 207.041 g/mol
Melting Pt: 179…181 °C
Storage Temperature: Ambient
MDL Number: MFCD05865213
CAS Number: 24287-95-4
Order Now
Specification Test Results
Appearance White to tan Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with HClO4 ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty