Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Butynyloxy)tetrahydro-2H-pyran 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42821-0050 42821-0250
TS42821-0050EA 93 USD InStock
TS42821-0050 TS42821-0250
2-(3-Butynyloxy)tetrahydro-2H-pyran 97%
2-(3-Butynyloxy)tetrahydro-2H-pyran


Formula: C₉H₁₄O₂
Boiling Pt: 92…95 °C (18 mmHg)
Density: 0.984 g/cm³
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012352
CAS Number: 40365-61-5
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty