Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethynylpyridine hydrochloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42849-0010 42849-0050
TS42849-0010EA 154 USD InStock
TS42849-0010 TS42849-0050
4-Ethynylpyridine hydrochloride 97%
4-Ethynylpyridine hydrochloride


Formula: C₇H₆ClN
Melting Pt: 150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093912
CAS Number: 352530-29-1
Order Now
Specification Test Results
Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
NMR consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty