Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylnicotinic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42942-0010 42942-0050
TS42942-0010EA 203 USD InStock
TS42942-0010 TS42942-0050
4-Methylnicotinic acid 97%
4-Methylnicotinic acid


Formula: C₇H₇NO₂
MW: 137.14
Melting Pt: 214…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00128869
CAS Number: 3222-50-2
Order Now
Specification Test Results
Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Melting point 212 to 220 °C
HPLC ≥96.0 %
Nitrate (NO3) ≤0.001 %
Phosphate (PO4) ≤0.0003 %
Aluminium (Al) ≤0.001 %
Arsenic (As) ≤0.00002 %
Cadmium (Cd) ≤0.0005 %
Calcium (Ca) ≤0.001 %
Chromium (Cr) ≤0.0005 %
Cobalt (Co) ≤0.0005 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.002 %
Lead (Pb) ≤0.0005 %
Magnesium (Mg) ≤0.0005 %
Manganese (Mn) ≤0.0005 %
Nickel (Ni) ≤0.0005 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.005 %
Zinc (Zn) ≤0.0005 %
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.005 % (in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty