Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-indazol-5-amine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42952-0010 42952-0050
TS42952-0010EA 121 USD InStock
TS42952-0010 TS42952-0050
1-Methyl-1H-indazol-5-amine 97%
1-Methyl-1H-indazol-5-amine


Formula: C₈H₉N₃
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD03305455
CAS Number: 50593-24-3
Order Now
Specification Test Results
Appearance Brown Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 154 °C
HPLC ≥97.0 %
UV 1 cm cell vs water
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity ≤0.0003 meq/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty