Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromothiazole-4-carbaldehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

42954-0010 42954-0050
TS42954-0010EA 121 USD InStock
TS42954-0010 TS42954-0050
2-Bromothiazole-4-carbaldehyde 97%
2-Bromothiazole-4-carbaldehyde


Formula: C₄H₂BrNOS
MW: 192.03 g/mol
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD04115726
CAS Number: 5198-80-1
Order Now
Specification Test Results
Appearance White to yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 130 °C
HPLC ≥96.0 %
UV at 250 nm A: ≤0.05
Non-volatile matter ≤0.0003 %
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty