Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzofurazan-5-boronic acid pinacol ester 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43047-0010 43047-0050
TS43047-0010EA 105 USD InStock
TS43047-0010 TS43047-0050
Benzofurazan-5-boronic acid pinacol ester 97%
Benzofurazan-5-boronic acid pinacol ester


Formula: C₁₂H₁₅BN₂O₃
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD09842717
CAS Number: 1073355-14-2
Order Now
Specification Test Results
Appearance White to yellow to tan Powder
Infrared spectrum Conforms
Melting point 86 to 91 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty