Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indazole-4-carbaldehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43103-0010 43103-0050
TS43103-0010EA 327 USD InStock
TS43103-0010 TS43103-0050
1H-Indazole-4-carbaldehyde 97%
1H-Indazole-4-carbaldehyde


Formula: C₈H₆N₂O
MW: 146.15 g/mol
Melting Pt: 166…167 °C
Storage Temperature: Ambient
MDL Number: MFCD06738279
CAS Number: 669050-70-8
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 165 to 169 °C
HPLC ≥96.0 %
Water adsorption ≥20 % (50% rH, 1 bar, 25 °C)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty