Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indazole-6-carboxaldehyde 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43104-0010 43104-0050
TS43104-0010EA 78 USD InStock
TS43104-0010 TS43104-0050
1H-Indazole-6-carboxaldehyde 97%
1H-Indazole-6-carboxaldehyde


Formula: C₈H₆N₂O
MW: 146.15 g/mol
Melting Pt: 180.6…180.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06738282
CAS Number: 669050-69-5
Order Now
Specification Test Results
Appearance White to yellow to brown Powder
Infrared spectrum Conforms
Melting point 180 to 185 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty