Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylsulfanylpyrimidine-4-carboxylic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43237-0010 43237-0050
TS43237-0010EA 170 USD InStock
TS43237-0010 TS43237-0050
2-Methylsulfanylpyrimidine-4-carboxylic acid 97%
2-Methylsulfanylpyrimidine-4-carboxylic acid


Formula: C₆H₆N₂O₂S
Melting Pt: 208…209 °C
Storage Temperature: Ambient
MDL Number: MFCD09834808
CAS Number: 1126-44-9
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty