Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-bromo-1H-indazole 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43576-0010 43576-0050
TS43576-0010EA 115 USD InStock
TS43576-0010 TS43576-0050
3-Amino-5-bromo-1H-indazole 95%
3-Amino-5-bromo-1H-indazole


Formula: C₇H₆BrN₃
Melting Pt: 237…238 °C
Storage Temperature: Ambient
MDL Number: MFCD03426696
CAS Number: 61272-71-7
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 174 to 178 °C
HPLC ≥94.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty