Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloropicolinic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43666-0010 43666-0050
TS43666-0010EA 36 USD InStock
TS43666-0010 TS43666-0050
6-Chloropicolinic acid 97%
6-Chloropicolinic acid


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: 162.6…162.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00155390
CAS Number: 4684-94-0
Order Now
Specification Test Results
Appearance White to cream to tan Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty