Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoropicolinic acid 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

43684-0010 43684-0050
TS43684-0010EA 227 USD InStock
TS43684-0010 TS43684-0050
6-Fluoropicolinic acid 97%
6-Fluoropicolinic acid


Formula: C₆H₄FNO₂
Melting Pt: 138.4…141.1 °C
Storage Temperature: Ambient
MDL Number: MFCD02181193
CAS Number: 402-69-7
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 135 to 142 °C
Water ≤1.0 %
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty